Assay Details
Target Gene Details
Entrez Gene ID: | 216188 |
Gene Name: | aldehyde dehydrogenase 1 family, member L2 |
Gene Aliases: |
D330038I09Rik, mtFDH |
Location: |
Chr.10:83487283-83534146 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aldh1l2 | NM_153543.2 | NP_705771.2 | ||
| AK052361.1 | ||||
| AK086413.1 | ||||
| AK141954.1 | ||||
| BC034531.1 | AAH34531.1 |
More Information
Set Membership: |
|

Genomic Map