Assay Details
Target Gene Details
Entrez Gene ID: | 216188 |
Gene Name: | aldehyde dehydrogenase 1 family, member L2 |
Gene Aliases: |
D330038I09Rik, mtFDH |
Location: |
Chr.10:83487283-83534146 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aldh1l2 | NM_153543.2 | 20 | 2498 | NP_705771.2 |
| AK052361.1 | 20 | 2389 | ||
| AK086413.1 | 19 | 2344 | ||
| AK141954.1 | 20 | 2459 | ||
| AK144594.1 | 2 | 216 | ||
| AK145046.1 | 1 | 44 | ||
| BC034531.1 | 20 | 2420 | AAH34531.1 |
More Information
Set Membership: |
|

Genomic Map