Assay Details
Target Gene Details
Entrez Gene ID: | 12952 |
Gene Name: | cryptochrome 1 (photolyase-like) |
Gene Aliases: |
AU020726, AU021000, Phll1 |
Location: |
Chr.10:85131700-85185054 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cry1 | NM_007771.3 | NP_031797.1 | ||
| AB000777.1 | BAA19175.1 | |||
| AF156986.1 | AAD39548.1 | |||
| AK133818.1 | ||||
| AK162460.1 | BAE36931.1 | |||
| BC022174.1 | AAH22174.1 | |||
| BC085499.1 | AAH85499.1 |
More Information
Set Membership: |
|

Genomic Map