Assay Details
Target Gene Details
Entrez Gene ID: | 77987 |
Gene Name: | activating signal cointegrator 1 complex subunit 3 |
Gene Aliases: |
ASC1p200, B630009I04Rik, BC023451, D430001L07Rik, D630041L21, Helic1, RNAH |
Location: |
Chr.10:50592669-50851204 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ascc3 | NM_198007.2 | 21 | 3638 | NP_932124.2 |
| BC059917.1 | 2 | 278 | AAH59917.1 |
More Information
Set Membership: |
|

Genomic Map