Assay Details
Target Gene Details
Entrez Gene ID: | 11540 |
Gene Name: | adenosine A2a receptor |
Gene Aliases: |
A2AAR, A2aR, AA2AR |
Location: |
Chr.10:75316860-75334792 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adora2a | NM_009630.3 | 1 | 117 | NP_033760.2 |
| BY168137.1 | 1 | 51 | ||
| BY349524.1 | 1 | 117 | ||
| CJ135665.1 | 1 | 51 |
More Information
Set Membership: |
|

Genomic Map