Assay Details
Target Gene Details
Entrez Gene ID: | 67884 |
Gene Name: | RIKEN cDNA 1810043G02 gene |
Gene Aliases: |
AV026620, D10Jhu13e |
Location: |
Chr.10:77978530-77985438 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1810043G02Rik | NM_026431.2 | NP_080707.2 | ||
| AK003097.1 | ||||
| AK004032.1 | BAB23134.1 | |||
| AK075794.1 | BAC35963.1 | |||
| AK147511.1 | 1 | 1124 | ||
| AK165293.1 | 2 | 162 | ||
| BC010330.1 | AAH10330.1 |

Genomic Map