Assay Details
Target Gene Details
Entrez Gene ID: | 67500 |
Gene Name: | cell division cycle and apoptosis regulator 1 |
Gene Aliases: |
2610511G16Rik, 9430036H15Rik, Carp1 |
Location: |
Chr.10:62743928-62792368 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccar1 | NM_026201.3 | 15 | 2078 | NP_080477.1 |
| AF465615.1 | 15 | 1969 | AAO17318.1 | |
| AK012111.1 | 15 | 1969 | BAB28040.2 | |
| AK031507.1 | 1 | 1720 | ||
| AK078424.1 | 1 | 55 | BAC37267.1 | |
| BC010199.1 | 15 | 1956 | ||
| BC034174.1 | 15 | 1979 | ||
| BC039939.1 | 15 | 1979 | AAH39939.1 | |
| BC060130.1 | 15 | 1966 | AAH60130.1 | |
| BC079652.1 | 15 | 1952 | AAH79652.1 | |
| DQ696475.1 |
More Information
Set Membership: |
|

Genomic Map