Assay Details
Target Gene Details
Entrez Gene ID: | 11846 |
Gene Name: | arginase, liver |
Gene Aliases: |
AI, AI256583, Arg-1, PGIF |
Location: |
Chr.10:24915207-24927470 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arg1 | NM_007482.3 | NP_031508.1 | ||
| AK149471.1 | BAE28901.1 | |||
| AK171417.1 | BAE42440.1 | |||
| BC013341.1 | AAH13341.1 | |||
| BC050005.1 | AAH50005.2 | |||
| CT010173.1 | CAJ18381.1 | |||
| U51805.1 | AAA98611.1 |
More Information
Set Membership: |
|

Genomic Map