Assay Details
Target Gene Details
Entrez Gene ID: | 67500 |
Gene Name: | cell division cycle and apoptosis regulator 1 |
Gene Aliases: |
2610511G16Rik, 9430036H15Rik, Carp1 |
Location: |
Chr.10:62743928-62792368 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccar1 | NM_026201.3 | 2 | 215 | NP_080477.1 |
| AF465615.1 | 2 | 106 | AAO17318.1 | |
| AK012111.1 | 2 | 106 | BAB28040.2 | |
| BC010199.1 | 2 | 93 | ||
| BC034174.1 | 2 | 116 | ||
| BC039939.1 | 2 | 115 | AAH39939.1 | |
| BC051052.1 | 2 | 117 | AAH51052.1 | |
| BC060130.1 | 2 | 103 | AAH60130.1 | |
| BC079652.1 | 2 | 89 | AAH79652.1 |
More Information
Set Membership: |
|

Genomic Map