Assay Details
Target Gene Details
Entrez Gene ID: | 67500 |
Gene Name: | cell division cycle and apoptosis regulator 1 |
Gene Aliases: |
2610511G16Rik, 9430036H15Rik, Carp1 |
Location: |
Chr.10:62743928-62792368 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 19 - Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccar1 | NM_026201.3 | NP_080477.1 | ||
| AF465615.1 | AAO17318.1 | |||
| AK077151.1 | BAC36646.1 | |||
| AK078424.1 | BAC37267.1 | |||
| BC079652.1 | AAH79652.1 |
More Information
Set Membership: |
|

Genomic Map