Assay Details
Target Gene Details
Entrez Gene ID: | 67500 |
Gene Name: | cell division cycle and apoptosis regulator 1 |
Gene Aliases: |
2610511G16Rik, 9430036H15Rik, Carp1 |
Location: |
Chr.10:62743928-62792368 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccar1 | NM_026201.3 | 6 | 613 | NP_080477.1 |
| AF465615.1 | 6 | 504 | AAO17318.1 | |
| AK012111.1 | 6 | 504 | BAB28040.2 | |
| AK215105.1 | ||||
| BC010199.1 | 6 | 491 | ||
| BC034174.1 | 6 | 514 | ||
| BC039939.1 | 6 | 514 | AAH39939.1 | |
| BC051052.1 | 6 | 515 | AAH51052.1 | |
| BC060130.1 | 6 | 501 | AAH60130.1 | |
| BC079652.1 | 6 | 487 | AAH79652.1 |
More Information
Set Membership: |
|

Genomic Map