Assay Details
Target Gene Details
Entrez Gene ID: | 74392 |
Gene Name: | sperm antigen with calponin homology and coiled-coil domains 1-like |
Gene Aliases: |
4930470P14Rik, 4932439K10Rik, 9530057A13Rik, AI317249, Cytsa, mKIAA0376 |
Location: |
Chr.10:75212073-75312400 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Specc1l | NM_001145826.1 | 6 | 689 | NP_001139298.1 |
| NM_153406.3 | 3 | 460 | NP_700455.3 | |
| AB093236.1 | 6 | 689 | BAC41420.1 | |
| AK035500.1 | 3 | 462 | ||
| AK052716.1 | 3 | 461 | BAC35113.1 | |
| AK172502.1 | 3 | 438 | ||
| AY884297.1 | 3 | 461 | AAX84188.1 | |
| BC072595.1 | 2 | 299 | AAH72595.1 |

Genomic Map