Assay Details
Target Gene Details
Entrez Gene ID: | 11537 |
Gene Name: | complement factor D (adipsin) |
Gene Aliases: |
Adn, DF |
Location: |
Chr.10:79890853-79892660 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cfd | NM_001291915.1 | NP_001278844.1 | ||
| NM_013459.3 | NP_038487.1 | |||
| AK143817.1 | BAE25550.1 | |||
| BC138778.1 | ||||
| BC138779.1 | ||||
| BC145412.1 |
More Information
Set Membership: |
|

Genomic Map