Assay Details
Target Gene Details
Entrez Gene ID: | 212862 |
Gene Name: | choline phosphotransferase 1 |
Gene Aliases: |
- |
Location: |
Chr.10:88472813-88503970 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chpt1 | NM_001146690.1 | NP_001140162.1 | ||
| NM_144807.3 | NP_659056.3 | |||
| NR_027477.1 | ||||
| AK007964.1 | ||||
| AK032497.1 | BAC27897.1 | |||
| AY445814.1 | AAR16089.1 | |||
| BE650891.1 | ||||
| EL958096.1 |
More Information
Set Membership: |
|

Genomic Map