Assay Details
Target Gene Details
Entrez Gene ID: | 56371 |
Gene Name: | fizzy/cell division cycle 20 related 1 (Drosophila) |
Gene Aliases: |
AW108046, Cdh1, FZR, FZR2, Fyr, HCDH, HCDH1 |
Location: |
Chr.10:81366879-81378370 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fzr1 | NM_019757.1 | 11 | 1353 | NP_062731.1 |
| AF083809.1 | 11 | 1353 | AAD52029.1 | |
| AK051993.1 | ||||
| AK081476.1 | 11 | 1409 | ||
| AK154822.1 | 11 | 1437 | BAE32854.1 | |
| AK155370.1 | 11 | 1435 | BAE33224.1 | |
| AK163951.1 | 11 | 1412 | BAE37547.1 | |
| AK172679.1 | 8 | 2373 | ||
| AK178744.1 | 2 | 275 | ||
| AK185038.1 | 1 | 74 | ||
| AK209977.1 | 2 | 275 | ||
| AK212079.1 | 1 | 74 | ||
| AK214012.1 | 1 | 74 | ||
| BC006616.1 | 11 | 1434 | AAH06616.1 | |
| FJ786633.2 | 6 | 587 |
More Information
Set Membership: |
|

Genomic Map