Assay Details
Target Gene Details
Entrez Gene ID: | 319880 |
Gene Name: | transmembrane and coiled coil domains 3 |
Gene Aliases: |
AW488095, C630016B22Rik, C88213, Tmcc1 |
Location: |
Chr.10:94311912-94590954 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmcc3 |

Genomic Map