Assay Details
Target Gene Details
Entrez Gene ID: | 14708 |
Gene Name: | guanine nucleotide binding protein (G protein), gamma 7 |
Gene Aliases: |
AI840417 |
Location: |
Chr.10:80948624-81014961 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gng7 | BY271138.1 |

Genomic Map