Assay Details
Target Gene Details
Entrez Gene ID: | 17179 |
Gene Name: | megakaryocyte-associated tyrosine kinase |
Gene Aliases: |
CHK, HYL, Ntk |
Location: |
Chr.10:81252795-81262982 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Matk | NM_001285853.1 | NP_001272782.1 | ||
| NM_001285854.1 | NP_001272783.1 | |||
| NM_001285855.1 | NP_001272784.1 | |||
| NM_010768.2 | NP_034898.1 | |||
| AK013606.1 | BAB28926.1 | |||
| AK079816.1 | ||||
| AK158883.1 | ||||
| BC111864.1 | AAI11865.1 | |||
| D45243.1 | BAA08199.1 | |||
| L27738.1 | AAB59677.1 | |||
| S77473.1 | AAB33566.1 | |||
| U05210.1 | AAA18829.1 |

Genomic Map