Assay Details
Target Gene Details
Entrez Gene ID: | 209462 |
Gene Name: | HECT domain and ankyrin repeat containing, E3 ubiquitin protein ligase 1 |
Gene Aliases: |
1700042J16Rik, A730034A22Rik, BC025474 |
Location: |
Chr.10:45577571-45712345 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 21 - Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hace1 | NM_172473.3 | NP_766061.2 | ||
| AK029162.1 | ||||
| AK042879.1 | BAC31390.1 | |||
| AK156958.1 | BAE33915.1 | |||
| AK220383.1 | BAD90440.1 | |||
| BC025227.1 | AAH25227.1 | |||
| BC025474.1 | AAH25474.1 | |||
| BC120695.1 | ||||
| BC120697.1 |
More Information
Set Membership: |
|

Genomic Map