Assay Details
Target Gene Details
Entrez Gene ID: | 268354 |
Gene Name: | family with sequence similarity 19, member A2 |
Gene Aliases: |
6330575M02, FAM19A2 TAFA-2, TAFA2, Tafa-2 |
Location: |
Chr.10:123264076-123741204 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam19a2 | NM_001252387.1 | NP_001239316.1 | ||
| NM_182807.3 | NP_877959.2 | |||
| NR_045514.1 | ||||
| AK032077.1 | ||||
| AK032483.1 | ||||
| AK139403.1 | BAE23995.1 | |||
| AK140763.1 | BAE24471.1 | |||
| AK164201.1 | BAE37679.1 | |||
| AY325121.1 | AAP92413.1 | |||
| BC027082.1 | AAH27082.1 |
More Information
Set Membership: |
|

Genomic Map