Assay Details
Target Gene Details
Entrez Gene ID: | 15364 |
Gene Name: | high mobility group AT-hook 2 |
Gene Aliases: |
9430083A20Rik, HMGI-C, Hmgic, pg, pygmy |
Location: |
Chr.10:120361275-120476935 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hmga2 | NM_010441.2 | NP_034571.1 | ||
| AK028346.1 | BAC25896.1 | |||
| AK035164.1 | 1 | 3039 | ||
| AK142059.1 | BAE24928.1 | |||
| BC052158.1 | AAH52158.1 | |||
| BC069985.1 | AAH69985.1 | |||
| BC085085.1 | AAH85085.1 | |||
| X58380.1 | CAA41270.1 |
More Information
Set Membership: |
|

Genomic Map