Assay Details
Target Gene Details
Entrez Gene ID: | 23943 |
Gene Name: | extended synaptotagmin-like protein 1 |
Gene Aliases: |
Fam62a, Mbc2, vp115 |
Location: |
Chr.10:128510248-128525859 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Esyt1 | NM_011843.2 | 17 | 1899 | NP_035973.1 |
| AF098634.1 | 8 | 692 | AAD10190.1 | |
| AK044656.1 | 17 | 1909 | BAC32020.1 | |
| AK054147.1 | 8 | 683 | ||
| AK080094.1 | 17 | 1912 | ||
| AK088421.1 | 17 | 1913 | ||
| AK142877.1 | 1 | 1270 | ||
| AK152482.1 | BAE31254.1 | |||
| AK152554.1 | BAE31308.1 | |||
| BC011482.1 | 17 | 1899 | AAH11482.1 |

Genomic Map