Assay Details
Target Gene Details
Entrez Gene ID: | 71567 |
Gene Name: | minichromosome maintenance complex component 9 |
Gene Aliases: |
9030408O17Rik, Gm235, Mcmdc1 |
Location: |
Chr.10:53536315-53630439 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mcm9 | NM_027830.2 | 10 | 1900 | NP_082106.2 |
| AK133212.1 | 3 | 313 | BAE21561.1 | |
| AK133345.1 | 3 | 1127 | ||
| BC062185.1 |
More Information
Set Membership: |
|

Genomic Map