Assay Details
Target Gene Details
Entrez Gene ID: | 23943 |
Gene Name: | extended synaptotagmin-like protein 1 |
Gene Aliases: |
Fam62a, Mbc2, vp115 |
Location: |
Chr.10:128510248-128525859 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Esyt1 | NM_011843.2 | 8 | 921 | NP_035973.1 |
| AK044656.1 | 8 | 931 | BAC32020.1 | |
| AK080094.1 | 8 | 934 | ||
| AK088421.1 | 8 | 934 | ||
| AK152482.1 | 8 | 934 | BAE31254.1 | |
| AK152554.1 | 8 | 934 | BAE31308.1 | |
| BC011482.1 | 8 | 921 | AAH11482.1 |

Genomic Map