Assay Details
Target Gene Details
Entrez Gene ID: | 23887 |
Gene Name: | gamma-glutamyltransferase 5 |
Gene Aliases: |
AI551243, GGL, GGT-REL, Ggtla1 |
Location: |
Chr.10:75589327-75616968 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ggt5 | NM_011820.4 | 5 | 964 | NP_035950.2 |
| AF077765.1 | 5 | 949 | AAC71001.1 | |
| AK049084.1 | 5 | 966 | ||
| AK052183.1 | 5 | 965 | BAC34873.1 | |
| AK085788.1 | 4 | 1446 | ||
| AK140308.1 | 4 | 1485 | ||
| AK170326.1 | 5 | 828 | BAE41720.1 | |
| BC051971.1 | AAH51971.1 | |||
| BC113775.1 | 5 | 688 |
More Information
Set Membership: |
|

Genomic Map