Assay Details
Target Gene Details
Entrez Gene ID: | 23887 |
Gene Name: | gamma-glutamyltransferase 5 |
Gene Aliases: |
AI551243, GGL, GGT-REL, Ggtla1 |
Location: |
Chr.10:75589327-75616968 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ggt5 | NM_011820.4 | NP_035950.2 | ||
| AF077765.1 | AAC71001.1 | |||
| AK029138.1 | BAC26318.1 | |||
| AK049084.1 | ||||
| AK052183.1 | BAC34873.1 | |||
| AK085788.1 | 5 | 2801 | ||
| AK140308.1 | ||||
| AK170326.1 | BAE41720.1 | |||
| BC051971.1 | AAH51971.1 | |||
| BC113775.1 |

Genomic Map