Assay Details
Target Gene Details
Entrez Gene ID: | 625249 |
Gene Name: | glutathione peroxidase 4 |
Gene Aliases: |
GPx-4, GSHPx-4, PHGPx, mtPHGPx, snGPx |
Location: |
Chr.10:80047166-80056439 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gpx4 | NM_001037741.3 | NP_001032830.2 | ||
| NM_008162.3 | NP_032188.3 | |||
| NR_110342.1 | ||||
| AB072797.1 | BAC87835.1 | |||
| AF045768.1 | AAC15832.1 | |||
| AF045769.1 | AAC15833.1 | |||
| AF274027.1 | AAK74112.1 | |||
| AK006441.1 | BAC55251.1 | |||
| AK145770.1 | ||||
| AK160389.1 | BAE35761.1 | |||
| AK160509.1 | BAE35833.1 | |||
| AK167881.1 | BAE39894.1 | |||
| AK168027.1 | BAE40012.1 | |||
| AK199684.1 | ||||
| BC083137.1 | ||||
| BC106147.1 | AAI06148.1 | |||
| BY238083.1 | ||||
| CA466677.1 | ||||
| D87896.1 | BAA22780.1 |
More Information
Set Membership: |
|

Genomic Map