Assay Details
Target Gene Details
Entrez Gene ID: | 18029 |
Gene Name: | nuclear factor I/C |
Gene Aliases: |
1110019L22Rik, 1500041O16Rik, AA589446, AI746521, NF1-C |
Location: |
Chr.10:81396191-81431167 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nfic | NM_008688.3 | 8 | 1251 | NP_032714.1 |
| NM_026756.2 | 8 | 1251 | NP_081032.2 | |
| AF358455.1 | 7 | 650 | AAK21331.1 | |
| AF358456.1 | 7 | 650 | AAK21332.1 | |
| AF358457.1 | 6 | 524 | AAK21333.1 | |
| AF358458.1 | AAK21334.1 | |||
| AF358459.1 | 5 | 266 | AAK21335.1 | |
| AF358460.1 | 5 | 266 | AAK21336.1 | |
| AK087539.1 | 8 | 1236 | BAC39918.1 | |
| AK147496.1 | 8 | 1236 | BAE27951.1 | |
| AK155258.1 | 8 | 1253 | BAE33151.1 | |
| U57635.1 | 8 | 1191 | AAB49930.1 | |
| Y07692.1 | 8 | 1210 | CAA68956.1 | |
| Y07693.1 | 8 | 1556 | CAA68957.1 |
More Information
Set Membership: |
|

Genomic Map