Assay Details
Target Gene Details
Entrez Gene ID: | 209558 |
Gene Name: | ectonucleotide pyrophosphatase/phosphodiesterase 3 |
Gene Aliases: |
AI876438, CD203c |
Location: |
Chr.10:24773814-24836195 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Enpp3 | NM_134005.2 | 9 | 925 | NP_598766.2 |
| AK132256.1 | 9 | 938 | ||
| AY630402.1 | 9 | 823 | AAT64421.1 | |
| BC117533.1 | 9 | 843 | ||
| BC118512.1 | 9 | 843 |

Genomic Map