Assay Details
Target Gene Details
Entrez Gene ID: | 16798 |
Gene Name: | large tumor suppressor |
Gene Aliases: |
AW208599 |
Location: |
Chr.10:7681209-7716461 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lats1 | NM_010690.1 | NP_034820.1 | ||
| AF104414.1 | AAD16883.1 | |||
| BC158092.1 | ||||
| BC158123.1 | ||||
| CO045800.1 |
More Information
Set Membership: |
|

Genomic Map