Assay Details
Target Gene Details
Entrez Gene ID: | 54427 |
Gene Name: | DNA (cytosine-5-)-methyltransferase 3-like |
Gene Aliases: |
D6Ertd14e, ecat7 |
Location: |
Chr.10:78042287-78063622 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 15 - Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dnmt3l | NM_001081695.2 | NP_001075164.1 | ||
| NM_001284197.1 | NP_001271126.1 | |||
| NM_001284198.1 | NP_001271127.1 | |||
| NM_001284199.1 | NP_001271128.1 | |||
| NM_001284200.1 | NP_001271129.1 | |||
| NM_019448.4 | NP_062321.1 | |||
| AF220524.1 | AAF73868.1 | |||
| AJ404467.1 | CAB94726.1 | |||
| AK010434.1 | BAB26936.1 | |||
| BC083147.1 | AAH83147.1 | |||
| EF051621.1 | ||||
| EF051622.1 | ||||
| EF051623.1 | ||||
| EF051624.1 |
More Information
Set Membership: |
|

Genomic Map