Assay Details
Target Gene Details
Entrez Gene ID: | 67074 |
Gene Name: | MON2 homolog (yeast) |
Gene Aliases: |
2610528O22Rik, AW495628, Sf21, mKIAA1040 |
Location: |
Chr.10:122992060-123076505 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mon2 | NM_001163024.1 | 14 | 2305 | NP_001156496.1 |
| NM_001163025.1 | 14 | 2308 | NP_001156497.1 | |
| NM_153395.2 | 14 | 2305 | NP_700444.2 | |
| AB014059.1 | 14 | 1827 | BAC11706.1 | |
| AK122425.1 | 14 | 2305 | BAC65707.1 | |
| AK163740.1 | 2 | 2149 | ||
| BC150943.1 | 14 | 1829 | ||
| CB244438.1 | 3 | 203 | ||
| CF747618.1 | 6 | 688 | ||
| CN525456.1 | 3 | 256 |
More Information
Set Membership: |
|

Genomic Map