Assay Details
Target Gene Details
Entrez Gene ID: | 18478 |
Gene Name: | phenylalanine hydroxylase |
Gene Aliases: |
AW106920 |
Location: |
Chr.10:87521795-87584137 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pah | NM_008777.3 | NP_032803.2 | ||
| AK145839.1 | ||||
| AK149519.1 | BAE28933.1 | |||
| BC013458.1 | AAH13458.1 | |||
| X51942.1 | CAA36205.1 |

Genomic Map