Assay Details
Target Gene Details
Entrez Gene ID: | 54427 |
Gene Name: | DNA (cytosine-5-)-methyltransferase 3-like |
Gene Aliases: |
D6Ertd14e, ecat7 |
Location: |
Chr.10:78042287-78063622 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dnmt3l | NM_001081695.2 | 3 | 349 | NP_001075164.1 |
| NM_001284197.1 | 1 | 190 | NP_001271126.1 | |
| NM_019448.4 | 2 | 203 | NP_062321.1 | |
| AF220524.1 | 1 | 134 | AAF73868.1 | |
| AJ404467.1 | 1 | 175 | CAB94726.1 | |
| AK010434.1 | 2 | 204 | BAB26936.1 | |
| AV473569.1 | 1 | 190 | ||
| BC083147.1 | 1 | 188 | AAH83147.1 | |
| EF051624.1 | 3 | 349 |
More Information
Set Membership: |
|

Genomic Map