Assay Details
Target Gene Details
Entrez Gene ID: | 14302 |
Gene Name: | fyn-related kinase |
Gene Aliases: |
BSK, BSK/IYK, C85044, GTK, RAK |
Location: |
Chr.10:34483400-34611534 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Frk | NM_001159544.1 | 1 | 139 | NP_001153016.1 |
| NM_010237.3 | 1 | 139 | NP_034367.2 | |
| AK028855.1 | 1 | 79 | BAC26155.1 | |
| AK052614.1 | BAC35063.1 | |||
| AK077264.1 | BAC36718.1 | |||
| AK085191.1 | ||||
| AK166379.1 | 1 | 139 | BAE38741.1 | |
| BC007137.1 | 1 | 152 | AAH07137.1 | |
| CN665865.1 | 1 | 139 | ||
| L36132.1 | AAA65197.1 | |||
| Z48757.1 | CAA88658.1 |

Genomic Map