Assay Details
Target Gene Details
Entrez Gene ID: | 117599 |
Gene Name: | helicase (DNA) B |
Gene Aliases: |
AI447783, D10Ertd664e |
Location: |
Chr.10:120083608-120112965 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Helb | NM_080446.2 | NP_536694.2 | ||
| AB048542.1 | BAB20809.1 | |||
| AK131122.1 | BAD21372.1 | |||
| BC068140.1 | AAH68140.1 |
More Information
Set Membership: |
|

Genomic Map