Assay Details
Target Gene Details
Entrez Gene ID: | 67074 |
Gene Name: | MON2 homolog (yeast) |
Gene Aliases: |
2610528O22Rik, AW495628, Sf21, mKIAA1040 |
Location: |
Chr.10:122992060-123076505 on Build GRCm38 |
Assay Gene Location: | Within Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mon2 | NM_001163024.1 | 26 | 3890 | NP_001156496.1 |
| NM_001163025.1 | 25 | 3890 | NP_001156497.1 | |
| NM_153395.2 | 25 | 3887 | NP_700444.2 | |
| AB014059.1 | 25 | 3409 | BAC11706.1 | |
| AK011826.1 | 4 | 678 | BAC25345.1 | |
| AK012182.2 | 5 | 827 | ||
| AK122425.1 | 26 | 3890 | BAC65707.1 | |
| AK162859.1 | 2 | 381 | BAE37087.1 | |
| BC150943.1 | 25 | 3411 | ||
| CN528444.1 | ||||
| CV563363.1 | 4 | 750 |
More Information
Set Membership: |
|

Genomic Map