Assay Details
Target Gene Details
Entrez Gene ID: | 18483 |
Gene Name: | paralemmin |
Gene Aliases: |
- |
Location: |
Chr.10:79793351-79820896 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Palm | NM_001161747.1 | NP_001155219.1 | ||
| NM_023128.4 | NP_075617.3 | |||
| AK037044.1 | BAC29683.1 | |||
| AK082163.1 | BAC38429.1 | |||
| AK133733.1 | BAE21809.1 | |||
| AK162411.1 | BAE36902.1 | |||
| BC015297.1 | AAH15297.1 | |||
| Y14771.1 | CAB37403.1 |
More Information
Set Membership: |
|

Genomic Map