Assay Details
Target Gene Details
Entrez Gene ID: | 83554 |
Gene Name: | follistatin-like 3 |
Gene Aliases: |
E030038F23Rik, Flrg |
Location: |
Chr.10:79777274-79782630 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fstl3 | NM_031380.2 | NP_113557.1 | ||
| AB024429.1 | BAB32663.1 | |||
| AF276238.1 | AAG47666.1 | |||
| AF310616.1 | AAG53666.1 | |||
| AK082898.1 | BAC38676.1 | |||
| AK087247.1 | BAC39827.1 | |||
| BC061052.1 | AAH61052.1 |
More Information
Set Membership: |
|

Genomic Map