Assay Details
Target Gene Details
Entrez Gene ID: | 83554 |
Gene Name: | follistatin-like 3 |
Gene Aliases: |
E030038F23Rik, Flrg |
Location: |
Chr.10:79777274-79782630 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fstl3 | NM_031380.2 | 4 | 557 | NP_113557.1 |
| AB024429.1 | 4 | 530 | BAB32663.1 | |
| AF276238.1 | 4 | 536 | AAG47666.1 | |
| AF310616.1 | 4 | 530 | AAG53666.1 | |
| AK079286.1 | 2 | 177 | ||
| AK082898.1 | 4 | 559 | BAC38676.1 | |
| AK087247.1 | 4 | 538 | BAC39827.1 | |
| BC061052.1 | 4 | 557 | AAH61052.1 |
More Information
Set Membership: |
|

Genomic Map