Assay Details
Target Gene Details
Entrez Gene ID: | 70208 |
Gene Name: | mediator complex subunit 23 |
Gene Aliases: |
130kDa, 3000002A17Rik, Crsp3, ESTM7, Sur2, X83317, mKIAA1216 |
Location: |
Chr.10:24869986-24913529 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Med23 | NM_001166416.1 | 22 | 2879 | NP_001159888.1 |
| NM_027347.3 | 21 | 2861 | NP_081623.3 | |
| AF507918.1 | 22 | 2829 | AAM28897.1 | |
| AK028545.1 | BAC26001.1 | |||
| AK129312.1 | 21 | 2794 | BAC98122.1 | |
| BC005508.1 | 5 | 724 | ||
| BC050916.1 | 21 | 2800 | AAH50916.1 |
More Information
Set Membership: |
|

Genomic Map