Assay Details
Target Gene Details
Entrez Gene ID: | 67345 |
Gene Name: | hect domain and RLD 4 |
Gene Aliases: |
1700056O17Rik, 4921531D01Rik, 9530080M15Rik, mKIAA1593 |
Location: |
Chr.10:63243782-63317881 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Herc4 | NM_026101.4 | NP_080377.2 | ||
| NM_030114.2 | NP_084390.1 | |||
| AK019546.1 | ||||
| AK143111.1 | BAE25272.1 | |||
| AK145735.1 | BAE26617.1 | |||
| AK147723.1 | BAE28095.1 | |||
| BC043082.1 | AAH43082.1 | |||
| BC060033.1 | AAH60033.1 |
More Information
Set Membership: |
|

Genomic Map