Assay Details
Target Gene Details
Entrez Gene ID: | 70208 |
Gene Name: | mediator complex subunit 23 |
Gene Aliases: |
130kDa, 3000002A17Rik, Crsp3, ESTM7, Sur2, X83317, mKIAA1216 |
Location: |
Chr.10:24869986-24913529 on Build GRCm38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Med23 | NM_001166416.1 | 24 | 3208 | NP_001159888.1 |
| NM_027347.3 | 23 | 3190 | NP_081623.3 | |
| AF507918.1 | 24 | 3158 | AAM28897.1 | |
| AK028545.1 | 24 | 3083 | BAC26001.1 | |
| AK129312.1 | 23 | 3123 | BAC98122.1 | |
| BC005508.1 | 7 | 1053 | ||
| BC050916.1 | 23 | 3129 | AAH50916.1 |
More Information
Set Membership: |
|

Genomic Map