Assay Details
Target Gene Details
Entrez Gene ID: | 237558 |
Gene Name: | myelin regulatory factor-like |
Gene Aliases: |
Gm239 |
Location: |
Chr.10:116776545-116896879 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Myrfl | NM_001033333.3 | 7 | 1010 | NP_001028505.1 |
| AK144383.1 | 7 | 1011 | BAE25858.1 | |
| AK144411.1 | 7 | 1011 | BAE25877.1 | |
| BC150915.1 | 7 | 814 | ||
| BC150923.1 | 7 | 814 |

Genomic Map