Assay Details
Target Gene Details
Entrez Gene ID: | 15273 |
Gene Name: | human immunodeficiency virus type I enhancer binding protein 2 |
Gene Aliases: |
MIBP1, Schnurri-2, Shn-2 |
Location: |
Chr.10:13965779-14151378 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hivep2 | NM_010437.2 | NP_034567.2 | ||
| AK137291.1 | BAE23294.1 | |||
| AK137384.1 | BAE23336.1 | |||
| AK147244.1 | BAE27792.1 | |||
| AK147348.1 | ||||
| AK147419.1 | BAE27900.1 | |||
| AK147655.1 | ||||
| BC150704.1 | ||||
| Y15907.1 | CAA75868.1 |

Genomic Map