Assay Details
Target Gene Details
Entrez Gene ID: | 19205 |
Gene Name: | polypyrimidine tract binding protein 1 |
Gene Aliases: |
AA407203, AL033359, HNRPI, PTB-1, PTB2, PTB3, PTB4, Ptb, pPTB |
Location: |
Chr.10:79854432-79864435 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |

Genomic Map