Assay Details
Target Gene Details
Entrez Gene ID: | 216152 |
Gene Name: | lipid phosphate phosphatase-related protein type 3 |
Gene Aliases: |
Prg2 |
Location: |
Chr.10:79860475-79874634 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lppr3 | NM_001170935.1 | NP_001164406.1 | ||
| BC098099.1 | AAH98099.1 |
Target Gene Details
Entrez Gene ID: | 19205 |
Gene Name: | polypyrimidine tract binding protein 1 |
Gene Aliases: |
AA407203, AL033359, HNRPI, PTB-1, PTB2, PTB3, PTB4, Ptb, pPTB |
Location: |
Chr.10:79854432-79864435 on Build GRCm38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptbp1 | NM_001077363.2 | NP_001070831.1 | ||
| NM_001283013.1 | NP_001269942.1 | |||
| NM_008956.3 | NP_032982.2 | |||
| AB074753.1 | BAB86943.1 | |||
| AB074764.1 | BAC65159.1 | |||
| AK033319.1 | BAC28230.1 | |||
| AK036745.1 | BAC29560.1 | |||
| AK037607.1 | ||||
| AK041143.1 | BAC30837.1 | |||
| AK043824.1 | BAC31665.1 | |||
| AK044956.1 | BAC32158.1 | |||
| AK050498.1 | BAC34292.1 | |||
| AK051669.1 | BAC34712.1 | |||
| AK053926.1 | ||||
| AK082600.1 | BAC38544.1 | |||
| AK088126.1 | BAC40161.1 | |||
| AK088483.1 | BAC40383.1 | |||
| AK149941.1 | BAE29183.1 | |||
| AK150840.1 | BAE29899.1 | |||
| AK151292.1 | BAE30277.1 | |||
| AK153561.1 | BAE32096.1 | |||
| AK163274.1 | BAE37269.1 | |||
| AK168359.1 | BAE40295.1 | |||
| AK168392.1 | ||||
| AK172712.1 | BAE43140.1 | |||
| AK206119.1 | ||||
| BC006666.1 | 1 | 402 | AAH06666.1 | |
| BC007472.1 | AAH07472.1 | |||
| BC028848.1 | AAH28848.1 | |||
| BC066210.1 | AAH66210.1 | |||
| BC086489.1 | AAH86489.1 | |||
| X52101.1 | CAA36321.1 |
More Information
Set Membership: |
|

Genomic Map