Assay Details
Target Gene Details
Entrez Gene ID: | 71137 |
Gene Name: | regulatory factor X, 4 (influences HLA class II expression) |
Gene Aliases: |
4933412G19Rik, NYD-sp10 |
Location: |
Chr.10:84756048-84906538 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rfx4 | NM_001024918.1 | 16 | 2132 | NP_001020089.1 |
| NM_027689.3 | 12 | 1464 | NP_081965.2 | |
| AB086957.1 | 16 | 1986 | BAD07408.1 | |
| AB089184.1 | 12 | 1465 | BAD07409.1 | |
| AK034131.1 | 16 | 2119 | BAC28598.1 | |
| AK132673.1 | 13 | 1527 | BAE21294.1 | |
| AY102010.2 | 16 | 2015 | AAM52485.2 | |
| AY342003.1 | 16 | 1997 | AAQ17042.1 | |
| BC138719.1 | 12 | 1452 | ||
| BC138720.1 | 12 | 1452 |
More Information
Set Membership: |
|

Genomic Map