Assay Details
Target Gene Details
Entrez Gene ID: | 18768 |
Gene Name: | protein kinase inhibitor beta, cAMP dependent, testis specific |
Gene Aliases: |
Pkibeta, Prkacn2 |
Location: |
Chr.10:57631981-57741112 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pkib | NM_001039051.2 | 1 | 91 | NP_001034140.1 |
| NM_008863.4 | 1 | 91 | NP_032889.3 | |
| AK005281.1 | ||||
| AK083875.1 | BAC39046.1 | |||
| AK089677.1 | ||||
| AK156851.1 | ||||
| AK156950.1 | BAE33910.1 | |||
| AK157521.1 | BAE34112.1 | |||
| AK171359.1 | ||||
| AK172041.1 | ||||
| L02241.1 | 1 | 98 | AAB59678.1 |
More Information
Set Membership: |
|

Genomic Map