Assay Details
Target Gene Details
Entrez Gene ID: | 216152 |
Gene Name: | lipid phosphate phosphatase-related protein type 3 |
Gene Aliases: |
Prg2 |
Location: |
Chr.10:79860475-79874634 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lppr3 | NM_001170935.1 | 9 | 2624 | NP_001164406.1 |
| BC098099.1 | 9 | 2377 | AAH98099.1 |
Target Gene Details
Entrez Gene ID: | 19205 |
Gene Name: | polypyrimidine tract binding protein 1 |
Gene Aliases: |
AA407203, AL033359, HNRPI, PTB-1, PTB2, PTB3, PTB4, Ptb, pPTB |
Location: |
Chr.10:79854432-79864435 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
More Information
Set Membership: |
|

Genomic Map